Product Details

SNP ID
rs144651140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27301877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAAATTGGACTGTTGCTTTTGCT[C/T]CAGATGGTTCATACTTTGCTTGGTC
Phenotype
MIM: 610091
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4522 PubMed Links

Gene Details

Gene
MIR4522
Gene Name
microRNA 4522
There are no transcripts associated with this gene.

Gene
WSB1
Gene Name
WD repeat and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015626.8 545 Missense Mutation CCA,TCA P44S NP_056441.6
NM_134265.2 545 Intron NP_599027.1
XM_005257963.3 545 Intron XP_005258020.1
XM_017024433.1 545 Missense Mutation CCA,TCA P44S XP_016879922.1
XM_017024434.1 545 UTR 5 XP_016879923.1
XM_017024435.1 545 UTR 5 XP_016879924.1
XM_017024436.1 545 Intron XP_016879925.1

View Full Product Details