Product Details

SNP ID
rs144673977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:413305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGACTCCTCCACCGTCAGCCTCCC[C/T]GACTTTGCCGAAATAGAGAACCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C17orf97 PubMed Links

Gene Details

Gene
C17orf97
Gene Name
chromosome 17 open reading frame 97
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013672.4 478 Silent Mutation CCC,CCT P154P NP_001013694.4
Gene
LOC105371430
Gene Name
uncharacterized LOC105371430
There are no transcripts associated with this gene.

View Full Product Details