Product Details

SNP ID
rs145180309
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:515207 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTAGCTGAATCACAGAGCTGTTA[C/G]CTGCTGCTTTCATCCACAGGTACTC
Phenotype
MIM: 615850
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
VPS53 PubMed Links

Gene Details

Gene
VPS53
Gene Name
VPS53, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128159.2 6808 UTR 3 NP_001121631.1
NM_018289.3 6808 Intron NP_060759.2
XM_011523953.2 6808 Intron XP_011522255.1
XM_017024817.1 6808 UTR 3 XP_016880306.1
XM_017024818.1 6808 UTR 3 XP_016880307.1

View Full Product Details