Product Details
- SNP ID
-
rs145539514
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:53823305 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGACTCTGCTGAACACCCCATGC[C/T]GCCCCCGCCCTTATCCCCCTTGGCT
- Phenotype
-
MIM: 615142
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIF2B
PubMed Links
Gene Details
- Gene
- KIF2B
- Gene Name
- kinesin family member 2B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032559.4 |
428 |
Missense Mutation |
CCG,CTG |
P91L |
NP_115948.4 |
View Full Product Details