Product Details

SNP ID
rs145621608
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7858460 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGCCAGGATATCAGCCACTGGT[C/T]TGATCCAAAGACCAGAGACGTGAGT
Phenotype
MIM: 611577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5D1 PubMed Links

Gene Details

Gene
CYB5D1
Gene Name
cytochrome b5 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005256440.3 386 Missense Mutation TCT,TTT S73F XP_005256497.1
XM_011523638.2 386 Missense Mutation TCT,TTT S73F XP_011521940.1
Gene
KDM6B
Gene Name
lysine demethylase 6B
There are no transcripts associated with this gene.

Gene
NAA38
Gene Name
N(alpha)-acetyltransferase 38, NatC auxiliary subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011524037.2 386 Intron XP_011522339.1
XM_017025225.1 386 Intron XP_016880714.1
XM_017025226.1 386 Intron XP_016880715.1
Gene
TMEM88
Gene Name
transmembrane protein 88
There are no transcripts associated with this gene.

View Full Product Details