Product Details

SNP ID
rs145931726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:79733624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGAGGTGGTGGGACAACGGCAGC[A/G]TGCCCATCTGGATCACAGCCCAGAG
Phenotype
MIM: 616997
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENPP7 PubMed Links

Gene Details

Gene
ENPP7
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178543.4 407 Missense Mutation ATG,GTG M124V NP_848638.3
XM_011524737.1 407 Missense Mutation ATG,GTG M197V XP_011523039.1

View Full Product Details