Product Details

SNP ID
rs146408284
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4953078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGAAGGCTGTGGAGAACATCA[A/G]CAATACTCTGGGCCCTGCTCTGCTG
Phenotype
MIM: 131370 MIM: 176610 MIM: 610431 MIM: 610056
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ENO3 PubMed Links

Gene Details

Gene
ENO3
Gene Name
enolase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193503.1 733 Intron NP_001180432.1
NM_001976.4 733 Intron NP_001967.3
NM_053013.3 733 Intron NP_443739.3
XM_005256521.2 733 Missense Mutation AAC,AGC N79S XP_005256578.1
XM_011523729.1 733 Missense Mutation AAC,AGC N70S XP_011522031.1
XM_017024346.1 733 Missense Mutation AAC,AGC N70S XP_016879835.1
Gene
PFN1
Gene Name
profilin 1
There are no transcripts associated with this gene.

Gene
RNF167
Gene Name
ring finger protein 167
There are no transcripts associated with this gene.

Gene
SPAG7
Gene Name
sperm associated antigen 7
There are no transcripts associated with this gene.

View Full Product Details