Product Details

SNP ID
rs146456630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40555228 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTGGATGGTGATAAAGGCCTC[C/T]ACATGCTCTGTGATGAGAGAGCATC
Phenotype
MIM: 600242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCR7 PubMed Links

Gene Details

Gene
CCR7
Gene Name
C-C motif chemokine receptor 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301714.1 732 Silent Mutation GTA,GTG V154V NP_001288643.1
NM_001301716.1 732 Silent Mutation GTA,GTG V211V NP_001288645.1
NM_001301717.1 732 Silent Mutation GTA,GTG V211V NP_001288646.1
NM_001301718.1 732 Silent Mutation GTA,GTG V211V NP_001288647.1
NM_001838.3 732 Silent Mutation GTA,GTG V217V NP_001829.1

View Full Product Details