Product Details

SNP ID
rs146578195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42160049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCTGGGCCAGGGTCAGTGGAA[C/T]GTGTCTGACCTGGACTGGAAACTGT
Phenotype
MIM: 604528 MIM: 604037
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNH4 PubMed Links

Gene Details

Gene
KCNH4
Gene Name
potassium voltage-gated channel subfamily H member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012285.2 3378 Silent Mutation ACA,ACG T1015T NP_036417.1
XM_017024400.1 3378 Silent Mutation ACA,ACG T1015T XP_016879889.1
XM_017024401.1 3378 Silent Mutation ACA,ACG T966T XP_016879890.1
XM_017024402.1 3378 Silent Mutation ACA,ACG T955T XP_016879891.1
XM_017024403.1 3378 Intron XP_016879892.1
Gene
RAB5C
Gene Name
RAB5C, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details