Product Details

SNP ID
rs146682384
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:2387078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGCCGTTGACTTGCTGGTGCGA[A/G]AGGGTGTGGGCGATGTGGCTCACTG
Phenotype
MIM: 603039 MIM: 611418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MNT PubMed Links

Gene Details

Gene
MNT
Gene Name
MAX network transcriptional repressor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020310.2 858 Silent Mutation CTC,CTT L524L NP_064706.1
XM_011523868.2 858 Silent Mutation CTC,CTT L532L XP_011522170.1
XM_011523869.1 858 Silent Mutation CTC,CTT L243L XP_011522171.1
XM_017024654.1 858 Silent Mutation CTC,CTT L243L XP_016880143.1
Gene
SGSM2
Gene Name
small G protein signaling modulator 2
There are no transcripts associated with this gene.

View Full Product Details