Product Details

SNP ID
rs146696963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18382571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCGGAGCTGGAGGAACAGATCG[C/T]GGAGCTCAACATCGTGCAGAAGGAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVPLL PubMed Links

Gene Details

Gene
EVPLL
Gene Name
envoplakin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145127.1 752 Silent Mutation CGC,CGT R135R NP_001138599.1
XM_011523982.2 752 Silent Mutation CGC,CGT R135R XP_011522284.1
XM_011523983.2 752 Silent Mutation CGC,CGT R135R XP_011522285.1
XM_011523985.2 752 Silent Mutation CGC,CGT R135R XP_011522287.1
XM_011523986.2 752 Intron XP_011522288.1
XM_011523987.2 752 Silent Mutation CGC,CGT R135R XP_011522289.1

View Full Product Details