Product Details

SNP ID
rs146794068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:27300119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACCTGTCATATGCCAGGTGCTAT[A/G]CTAGCTGTTGATGGAGCATACATTC
Phenotype
MIM: 610091
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR4522 PubMed Links

Gene Details

Gene
MIR4522
Gene Name
microRNA 4522
There are no transcripts associated with this gene.

Gene
WSB1
Gene Name
WD repeat and SOCS box containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015626.8 Intron NP_056441.6
NM_134265.2 Intron NP_599027.1
XM_005257963.3 Intron XP_005258020.1
XM_017024433.1 Intron XP_016879922.1
XM_017024434.1 Intron XP_016879923.1
XM_017024435.1 Intron XP_016879924.1
XM_017024436.1 Intron XP_016879925.1

View Full Product Details