Product Details

SNP ID
rs146982096
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35740498 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCTGCTTTGACAGCTTTGAGTA[C/T]GTCAAGACCATCCGCCAGCAGATCC
Phenotype
MIM: 611398 MIM: 612128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GAS2L2 PubMed Links

Gene Details

Gene
GAS2L2
Gene Name
growth arrest specific 2 like 2
There are no transcripts associated with this gene.

Gene
RASL10B
Gene Name
RAS like family 10 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033315.3 672 Silent Mutation TAC,TAT Y102Y NP_201572.1
XM_017025300.1 672 Silent Mutation TAC,TAT Y161Y XP_016880789.1
XM_017025301.1 672 Silent Mutation TAC,TAT Y102Y XP_016880790.1

View Full Product Details