Product Details

SNP ID
rs147361615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74843272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCGGTCCCCAGCCCGTGGGGCT[C/T]GGCTCTGGGGGCGGGTCGGGGGTGG
Phenotype
MIM: 138254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GRIN2C PubMed Links

Gene Details

Gene
GRIN2C
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000835.4 4312 Silent Mutation CCA,CCG P955P NP_000826.2
NM_001278553.1 4312 Intron NP_001265482.1
XM_006721845.3 4312 Silent Mutation CCA,CCG P1030P XP_006721908.1
XM_006721846.3 4312 Silent Mutation CCA,CCG P1029P XP_006721909.1
XM_011524686.2 4312 Silent Mutation CCA,CCG P1058P XP_011522988.1
XM_011524687.2 4312 Silent Mutation CCA,CCG P1051P XP_011522989.1
XM_011524688.2 4312 Silent Mutation CCA,CCG P1038P XP_011522990.1
XM_011524689.2 4312 Silent Mutation CCA,CCG P984P XP_011522991.1
XM_011524692.2 4312 Intron XP_011522994.1
XM_017024544.1 4312 Silent Mutation CCA,CCG P1059P XP_016880033.1
Gene
TMEM104
Gene Name
transmembrane protein 104
There are no transcripts associated with this gene.

View Full Product Details