Product Details
- SNP ID
-
rs147552949
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:63704562 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCCAACAGGCAGGGCACTGTGCC[A/G]TTCAGTTTCTCTAGCAGCATCTGGG
- Phenotype
-
MIM: 602539
MIM: 608626
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LIMD2
PubMed Links
Gene Details
- Gene
- LIMD2
- Gene Name
- LIM domain containing 2
There are no transcripts associated with this gene.
- Gene
- LOC729683
- Gene Name
- uncharacterized LOC729683
There are no transcripts associated with this gene.
- Gene
- MAP3K3
- Gene Name
- mitogen-activated protein kinase kinase kinase 3
There are no transcripts associated with this gene.
- Gene
- STRADA
- Gene Name
- STE20-related kinase adaptor alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001003786.2 |
975 |
Silent Mutation |
AAC,AAT |
N256N |
NP_001003786.1 |
NM_001003787.2 |
975 |
Silent Mutation |
AAC,AAT |
N293N |
NP_001003787.1 |
NM_001003788.2 |
975 |
Silent Mutation |
AAC,AAT |
N235N |
NP_001003788.1 |
NM_001165969.1 |
975 |
Silent Mutation |
AAC,AAT |
N264N |
NP_001159441.1 |
NM_001165970.1 |
975 |
Silent Mutation |
AAC,AAT |
N249N |
NP_001159442.1 |
NM_153335.5 |
975 |
Silent Mutation |
AAC,AAT |
N256N |
NP_699166.2 |
XM_005257797.2 |
975 |
Silent Mutation |
AAC,AAT |
N285N |
XP_005257854.1 |
XM_005257798.2 |
975 |
Silent Mutation |
AAC,AAT |
N264N |
XP_005257855.1 |
XM_005257799.2 |
975 |
Silent Mutation |
AAC,AAT |
N235N |
XP_005257856.1 |
XM_005257800.2 |
975 |
Silent Mutation |
AAC,AAT |
N293N |
XP_005257857.1 |
XM_005257801.4 |
975 |
UTR 3 |
|
|
XP_005257858.1 |
XM_005257803.4 |
975 |
UTR 3 |
|
|
XP_005257860.1 |
XM_011525466.2 |
975 |
Silent Mutation |
AAC,AAT |
N293N |
XP_011523768.1 |
XM_011525467.2 |
975 |
Silent Mutation |
AAC,AAT |
N258N |
XP_011523769.1 |
XM_017025312.1 |
975 |
Silent Mutation |
AAC,AAT |
N256N |
XP_016880801.1 |
XM_017025313.1 |
975 |
Silent Mutation |
AAC,AAT |
N235N |
XP_016880802.1 |
XM_017025314.1 |
975 |
UTR 3 |
|
|
XP_016880803.1 |
View Full Product Details