Product Details
- SNP ID
-
rs147570224
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:82741376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCGGCTTCCACACGGTGACGTGC[C/T]GCGGCTTCATCCCGCAGGTGAGTGC
- Phenotype
-
MIM: 608425
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FN3K
PubMed Links
Gene Details
- Gene
- FN3K
- Gene Name
- fructosamine 3 kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022158.3 |
512 |
Missense Mutation |
CGC,TGC |
R151C |
NP_071441.1 |
View Full Product Details