Product Details

SNP ID
rs147691445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7931787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCCATTCGCTGTAGTGCAGAC[A/C]CACTCCATTCCGGTCAAACAGGTAC
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037144.5 103 Intron NP_001032221.1
NM_053051.3 103 Intron NP_444279.2
XM_005256437.2 103 Intron XP_005256494.1
XM_005256438.2 103 Intron XP_005256495.1
XM_005256439.2 103 Intron XP_005256496.1
XM_017024128.1 103 Intron XP_016879617.1
XM_017024129.1 103 Intron XP_016879618.1
XM_017024130.1 103 Intron XP_016879619.1
XM_017024131.1 103 Intron XP_016879620.1
XM_017024132.1 103 Intron XP_016879621.1
XM_017024133.1 103 Intron XP_016879622.1
XM_017024134.1 103 Intron XP_016879623.1
XM_017024135.1 103 Intron XP_016879624.1
XM_017024136.1 103 Intron XP_016879625.1
XM_017024137.1 103 Intron XP_016879626.1
XM_017024138.1 103 Intron XP_016879627.1
XM_017024139.1 103 Intron XP_016879628.1
XM_017024140.1 103 Intron XP_016879629.1
XM_017024141.1 103 Intron XP_016879630.1
XM_017024142.1 103 Intron XP_016879631.1
XM_017024143.1 103 Intron XP_016879632.1
XM_017024144.1 103 Intron XP_016879633.1
Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166621.1 103 Missense Mutation NP_001160093.1
NM_021210.4 103 Missense Mutation NP_067033.1

View Full Product Details