Product Details

SNP ID
hCV164717478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68274014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCCGGTGCTGACAAGTAGCCCCC[C/G]CAACATCACTACCAGACGGTGTCCG
Phenotype
MIM: 610008 MIM: 603880
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARSG PubMed Links

Gene Details

Gene
ARSG
Gene Name
arylsulfatase G
There are no transcripts associated with this gene.

Gene
SLC16A6
Gene Name
solute carrier family 16 member 6
There are no transcripts associated with this gene.

View Full Product Details