Product Details

SNP ID
rs147973494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9772876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATCGTATTCCTCCACTTCGGC[A/T]CGGAGGCAGTCAAAGAAGCCCAGGG
Phenotype
MIM: 616161
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DHRS7C PubMed Links

Gene Details

Gene
DHRS7C
Gene Name
dehydrogenase/reductase 7C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105571.2 631 Silent Mutation CGA,CGT R206R NP_001099041.1
NM_001220493.1 631 Silent Mutation CGA,CGT R207R NP_001207422.1

View Full Product Details