Product Details
- SNP ID
-
rs148036764
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:7289617 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCGCTGGAAGTAGGGGCGGTTTCG[G/T]GGGCGCTGGGGCTCAGGCCGGGAAC
- Phenotype
-
MIM: 138190
MIM: 611447
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC2A4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3833134,rs55781423] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC2A4
- Gene Name
- solute carrier family 2 member 4
There are no transcripts associated with this gene.
- Gene
- YBX2
- Gene Name
- Y-box binding protein 2
View Full Product Details