Product Details

SNP ID
rs148321092
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:68042912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGCCTTTTTTTTCAGGTGATGG[A/C]TCAGTGTTCCGAGACTTGGTTATTA
Phenotype
MIM: 600685
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KPNA2 PubMed Links

Gene Details

Gene
KPNA2
Gene Name
karyopherin subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320611.1 1199 Silent Mutation GGA,GGC G193G NP_001307540.1
NM_002266.3 1199 Intron NP_002257.1

View Full Product Details