Product Details

SNP ID
rs148354373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:61412033 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGGATTTATACCCCGAGCCT[G/T]CCTTCCTCCGCTCTGGGAAGGACCC
Phenotype
MIM: 600747
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C17orf82 PubMed Links

Gene Details

Gene
C17orf82
Gene Name
chromosome 17 open reading frame 82
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203425.2 283 Missense Mutation GCC,TCC A20S NP_982249.2
Gene
TBX2
Gene Name
T-box 2
There are no transcripts associated with this gene.

View Full Product Details