Product Details

SNP ID
rs148601715
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:38326745 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGCCCATGGACTAAAAGGCAT[A/G]GTGGTTTTTCAGGAGCTGTGGGGAA
Phenotype
MIM: 614515 MIM: 611850
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR179 PubMed Links

Gene Details

Gene
GPR179
Gene Name
G protein-coupled receptor 179
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004334.3 6845 Missense Mutation NP_001004334.3
Gene
MRPL45
Gene Name
mitochondrial ribosomal protein L45
There are no transcripts associated with this gene.

View Full Product Details