Product Details

SNP ID
rs148627254
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74844241 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACTTATCTGGGTAGGTGCCCTTAA[A/G]ACTTTGGCCTGTGTGGGGAGGGGTG
Phenotype
MIM: 138254
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIN2C PubMed Links

Gene Details

Gene
GRIN2C
Gene Name
glutamate ionotropic receptor NMDA type subunit 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000835.4 3028 Intron NP_000826.2
NM_001278553.1 3028 Missense Mutation TCT,TTT S873F NP_001265482.1
XM_006721845.3 3028 Intron XP_006721908.1
XM_006721846.3 3028 Intron XP_006721909.1
XM_011524686.2 3028 Intron XP_011522988.1
XM_011524687.2 3028 Intron XP_011522989.1
XM_011524688.2 3028 Intron XP_011522990.1
XM_011524689.2 3028 Intron XP_011522991.1
XM_011524692.2 3028 Intron XP_011522994.1
XM_017024544.1 3028 Intron XP_016880033.1
Gene
TMEM104
Gene Name
transmembrane protein 104
There are no transcripts associated with this gene.

View Full Product Details