Product Details

SNP ID
rs148662684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44850361 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTACACAATGTACAGCGATTAC[A/G]TCAAGAGGATGGCACAGGATGCTGG
Phenotype
MIM: 603892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFTUD2 PubMed Links

Gene Details

Gene
EFTUD2
Gene Name
elongation factor Tu GTP binding domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142605.1 4230 UTR 3 NP_001136077.1
NM_001258353.1 4230 UTR 3 NP_001245282.1
NM_001258354.1 4230 UTR 3 NP_001245283.1
NM_004247.3 4230 UTR 3 NP_004238.3
Gene
HIGD1B
Gene Name
HIG1 hypoxia inducible domain family member 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271880.1 4230 Missense Mutation ATC,GTC I89V NP_001258809.1
NM_016438.3 4230 Missense Mutation ATC,GTC I89V NP_057522.1
XM_006721946.2 4230 Missense Mutation ATC,GTC I89V XP_006722009.1
XM_006721947.3 4230 Missense Mutation ATC,GTC I89V XP_006722010.1
XM_006721948.3 4230 Missense Mutation ATC,GTC I89V XP_006722011.1
XM_011524891.2 4230 Missense Mutation ATC,GTC I89V XP_011523193.1
XM_017024742.1 4230 Missense Mutation ATC,GTC I89V XP_016880231.1
XM_017024743.1 4230 Missense Mutation ATC,GTC I89V XP_016880232.1

View Full Product Details