Product Details
- SNP ID
-
rs148928902
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40777039 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTTCATTCTTGTTGTTGACTTTG[A/G]TGGATTTCTCTTCCACAGTGTGAAC
- Phenotype
-
MIM: 616675
MIM: 616676
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KRT26
PubMed Links
Gene Details
- Gene
- KRT26
- Gene Name
- keratin 26
There are no transcripts associated with this gene.
- Gene
- KRT27
- Gene Name
- keratin 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181537.3 |
1381 |
Missense Mutation |
ACC,ATC |
T447I |
NP_853515.2 |
View Full Product Details