Product Details

SNP ID
rs148934120
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31856807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCCATCTACTTGCCTAGTCGA[G/T]CCATAGCCTTCTCAGTCTCTCTCTC
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
COPRS PubMed Links

Gene Details

Gene
COPRS
Gene Name
coordinator of PRMT5 and differentiation stimulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018405.3 266 Missense Mutation GAT,GCT D53A NP_060875.2
XM_005258000.1 266 Missense Mutation GAT,GCT D41A XP_005258057.1
Gene
UTP6
Gene Name
UTP6, small subunit processome component
There are no transcripts associated with this gene.

View Full Product Details