Product Details

SNP ID
rs149151417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7923791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGACTTTGGCTTGTTGCTTCTTG[C/G]CATCTTCACTCTGCACTTTGTCCTT
Phenotype
MIM: 611425 MIM: 604111 MIM: 610969
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNTROB PubMed Links

Gene Details

Gene
CNTROB
Gene Name
centrobin, centriole duplication and spindle assembly protein
There are no transcripts associated with this gene.

Gene
KCNAB3
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004732.3 915 Missense Mutation GCC,GGC A323G NP_004723.2
XM_011524068.1 915 Missense Mutation GCC,GGC A274G XP_011522370.1
XM_017025304.1 915 Missense Mutation GCC,GGC A274G XP_016880793.1
XM_017025305.1 915 Missense Mutation GCC,GGC A245G XP_016880794.1
XM_017025306.1 915 Missense Mutation GCC,GGC A245G XP_016880795.1
Gene
LOC284023
Gene Name
uncharacterized LOC284023
There are no transcripts associated with this gene.

Gene
TRAPPC1
Gene Name
trafficking protein particle complex 1
There are no transcripts associated with this gene.

View Full Product Details