Product Details

SNP ID
rs149237363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57258303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTCGTCACGTTCGCAGACCCAGC[A/G]AGTGTAGATAAAGTATTAGGTCAGC
Phenotype
MIM: 607897
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSI2 PubMed Links

Gene Details

Gene
MSI2
Gene Name
musashi RNA binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322250.1 259 Silent Mutation GCA,GCG A51A NP_001309179.1
NM_001322251.1 259 Silent Mutation GCA,GCG A73A NP_001309180.1
NM_138962.3 259 Silent Mutation GCA,GCG A73A NP_620412.1
NM_170721.1 259 Silent Mutation GCA,GCG A69A NP_733839.1
XM_005257014.3 259 Silent Mutation GCA,GCG A73A XP_005257071.1
XM_005257015.3 259 Silent Mutation GCA,GCG A73A XP_005257072.1
XM_011524286.1 259 UTR 5 XP_011522588.1
XM_011524287.2 259 Intron XP_011522589.1
XM_017024147.1 259 Silent Mutation GCA,GCG A73A XP_016879636.1
XM_017024148.1 259 Silent Mutation GCA,GCG A73A XP_016879637.1
XM_017024149.1 259 Intron XP_016879638.1

View Full Product Details