Product Details

SNP ID
rs149378331
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:63746904 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTACATGGCTTTCACTTTGATTT[A/G]TTTCATTTTCATTTGCTTCTTTTCC
Phenotype
MIM: 608626
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC47 PubMed Links

Gene Details

Gene
CCDC47
Gene Name
coiled-coil domain containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020198.2 1633 Nonsense Mutation CAA,TAA Q477* NP_064583.2
XM_005257527.2 1633 Nonsense Mutation CAA,TAA Q477* XP_005257584.1
Gene
STRADA
Gene Name
STE20-related kinase adaptor alpha
There are no transcripts associated with this gene.

View Full Product Details