Product Details

SNP ID
rs149380490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35130371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCAAGGAACCCCGGCAAAAGAT[C/T]GTGTCCATCGGGCCGGAGGAGATGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FNDC8 PubMed Links

Gene Details

Gene
FNDC8
Gene Name
fibronectin type III domain containing 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017559.2 994 Silent Mutation ATC,ATT I304I NP_060029.1
Gene
NLE1
Gene Name
notchless homolog 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014445.2 994 Intron NP_001014445.1
NM_018096.4 994 Intron NP_060566.2
XM_017024777.1 994 UTR 3 XP_016880266.1
Gene
RAD51L3-RFFL
Gene Name
RAD51L3-RFFL readthrough
There are no transcripts associated with this gene.

View Full Product Details