Product Details

SNP ID
rs149778906
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:41168139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGACCTTGACCACAGCTCTGGT[A/C]AGAGCAGACAGAGCCACAGCAGGAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KRTAP4-2 PubMed Links

Gene Details

Gene
KRTAP4-2
Gene Name
keratin associated protein 4-2
There are no transcripts associated with this gene.

Gene
KRTAP4-3
Gene Name
keratin associated protein 4-3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033187.1 34 Missense Mutation GAC,TAC D12Y NP_149443.1
Gene
KRTAP4-4
Gene Name
keratin associated protein 4-4
There are no transcripts associated with this gene.

View Full Product Details