Product Details

SNP ID
rs149814069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:57869181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCATCTTCAGACACAGCGGGGTTG[A/G]CGTCGCCAGTTCCTGGAAGGAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CUEDC1 PubMed Links

Gene Details

Gene
CUEDC1
Gene Name
CUE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271875.1 683 Missense Mutation GCC,GTC A294V NP_001258804.1
NM_001292025.1 683 Missense Mutation GCC,GTC A294V NP_001278954.1
XM_006721900.2 683 Missense Mutation GCC,GTC A294V XP_006721963.1
XM_011524812.2 683 Missense Mutation GCC,GTC A187V XP_011523114.1
XM_017024640.1 683 Missense Mutation GCC,GTC A266V XP_016880129.1
XM_017024641.1 683 Missense Mutation GCC,GTC A212V XP_016880130.1

View Full Product Details