Product Details

SNP ID
rs149833913
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:31937457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCGGCGGGGGCTGCGGTGTTACCG[A/G]TGAAGAAGCCGAAAATGGAGCACGT
Phenotype
MIM: 606245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SUZ12 PubMed Links

Gene Details

Gene
SUZ12
Gene Name
SUZ12 polycomb repressive complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321207.1 448 Missense Mutation ATG,GTG M71V NP_001308136.1
NM_015355.3 448 Missense Mutation ATG,GTG M71V NP_056170.2
XM_006721794.3 448 UTR 5 XP_006721857.1
XM_017024409.1 448 UTR 5 XP_016879898.1

View Full Product Details