Product Details

SNP ID
rs150043620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7283770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCATGCTGGTCAACAATGTCCTGG[C/T]GGTGCTGGGGGGCAGCCTCATGGGC
Phenotype
MIM: 138190 MIM: 611447
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC2A4 PubMed Links

Gene Details

Gene
SLC2A4
Gene Name
solute carrier family 2 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042.2 556 Missense Mutation GCG,GTG A119V NP_001033.1
Gene
YBX2
Gene Name
Y-box binding protein 2
There are no transcripts associated with this gene.

View Full Product Details