Product Details

SNP ID
rs150314171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1769866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGCCAGGGCTCCCCAGACCC[C/T]GACAGCACAGGGGCGCTGGTGGAGG
Phenotype
MIM: 172860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERPINF1 PubMed Links

Gene Details

Gene
SERPINF1
Gene Name
serpin family F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017024754.1 176 Silent Mutation CCC,CCT P33P XP_016880243.1
Gene
SMYD4
Gene Name
SET and MYND domain containing 4
There are no transcripts associated with this gene.

View Full Product Details