Product Details

SNP ID
rs150727776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10825264 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAGGAAGAAGGACTTGAGGCTGC[A/G]TAAGAAATTCGACTTCTGTCTGTGC
Phenotype
MIM: 612068
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIRT PubMed Links

Gene Details

Gene
PIRT
Gene Name
phosphoinositide interacting regulator of transient receptor potential channels
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101387.1 677 Missense Mutation CGC,TGC R128C NP_001094857.1

View Full Product Details