Product Details

SNP ID
rs150976338
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7560782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGTCATAATTCCCCGGGCAAGGGC[A/G]AAACTTAACCTCTCTCCACATGGAA
Phenotype
MIM: 612844 MIM: 602695 MIM: 604472
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SENP3 PubMed Links

Gene Details

Gene
SENP3
Gene Name
SUMO1/sentrin/SMT3 specific peptidase 3
There are no transcripts associated with this gene.

Gene
SENP3-EIF4A1
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
TNFSF12
Gene Name
tumor necrosis factor superfamily member 12
There are no transcripts associated with this gene.

Gene
TNFSF12-TNFSF13
Gene Name
TNFSF12-TNFSF13 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172089.3 1450 Silent Mutation GCA,GCG A314A NP_742086.1
Gene
TNFSF13
Gene Name
tumor necrosis factor superfamily member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198622.1 1450 Silent Mutation GCA,GCG A207A NP_001185551.1
NM_001198623.1 1450 Silent Mutation GCA,GCG A206A NP_001185552.1
NM_001198624.1 1450 Silent Mutation GCA,GCG A189A NP_001185553.1
NM_003808.3 1450 Silent Mutation GCA,GCG A234A NP_003799.1
NM_172087.2 1450 Silent Mutation GCA,GCG A218A NP_742084.1
NM_172088.2 1450 Silent Mutation GCA,GCG A234A NP_742085.1

View Full Product Details