Product Details
- SNP ID
-
rs151274156
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:49991550 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCCCAGGGTTGGGCGGGGGCCC[A/G]GGAGAGGCATGGTGCAGGGTGGCTG
- Phenotype
-
MIM: 600525
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DLX3
PubMed Links
Gene Details
- Gene
- DLX3
- Gene Name
- distal-less homeobox 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005220.2 |
1057 |
Silent Mutation |
CCC,CCT |
P277P |
NP_005211.1 |
View Full Product Details