Product Details

SNP ID
rs138590264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63637924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGTGTAGGACTTTAGATACTGAG[A/G]GACCGTGGCTCCAGGTCATGCCTGA
Phenotype
MIM: 600518
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB4 PubMed Links

Gene Details

Gene
SERPINB4
Gene Name
serpin family B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002974.3 1115 Missense Mutation CCC,CTC P323L NP_002965.1
NM_175041.1 1115 Missense Mutation CCC,CTC P302L NP_778206.1
XM_011526138.1 1115 Missense Mutation CCC,CTC P323L XP_011524440.1

View Full Product Details