Product Details

SNP ID
rs138598889
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:42746299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATTTTAAAAGAATAATTCAAC[C/T]GACCACAGTCAATATAAATGCATTA
Phenotype
MIM: 609592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RIT2 PubMed Links

Gene Details

Gene
RIT2
Gene Name
Ras like without CAAX 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272077.1 Intron NP_001259006.1
NM_002930.3 Intron NP_002921.1

View Full Product Details