Product Details

SNP ID
rs141213977
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:48921471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTTCACAAAGCTGATCTGCAC[A/G]GTAAAGCCCGTCCACGGCTGCTGCA
Phenotype
MIM: 602932
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMAD7 PubMed Links

Gene Details

Gene
SMAD7
Gene Name
SMAD family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190821.1 767 Silent Mutation NP_001177750.1
NM_001190822.1 767 Silent Mutation NP_001177751.1
NM_001190823.1 767 Silent Mutation NP_001177752.1
NM_005904.3 767 Silent Mutation NP_005895.1

View Full Product Details