Product Details

SNP ID
rs142364104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:10672769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGTTCCAGGGCCTGAGAGTTCG[C/G]ATTGTATATTCTGTTTCCAGTCAGG
Phenotype
MIM: 613629
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101927410 PubMed Links

Gene Details

Gene
LOC101927410
Gene Name
uncharacterized LOC101927410
There are no transcripts associated with this gene.

Gene
PIEZO2
Gene Name
piezo type mechanosensitive ion channel component 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022068.3 9031 Missense Mutation CCG,GCG P2643A NP_071351.2
XM_011525723.2 9031 Missense Mutation CCG,GCG P2687A XP_011524025.1
XM_011525724.2 9031 Missense Mutation CCG,GCG P2668A XP_011524026.1
XM_011525725.2 9031 Missense Mutation CCG,GCG P2657A XP_011524027.1
XM_011525726.2 9031 Missense Mutation CCG,GCG P2626A XP_011524028.1
XM_017025918.1 9031 Missense Mutation CCG,GCG P2674A XP_016881407.1

View Full Product Details