Product Details

SNP ID
rs145104474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63639761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGCCAATAGTCCCATCAGGAAAT[A/G]GGTTTTTAATTTTTTCTGCAAGGGA
Phenotype
MIM: 600518
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINB4 PubMed Links

Gene Details

Gene
SERPINB4
Gene Name
serpin family B member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002974.3 632 Missense Mutation CCA,CTA P162L NP_002965.1
NM_175041.1 632 Missense Mutation CCA,CTA P162L NP_778206.1
XM_011526138.1 632 Missense Mutation CCA,CTA P162L XP_011524440.1

View Full Product Details