Product Details

SNP ID
rs148490056
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:75195195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCATTAAAACTAAATTATGGATA[G/T]TAAGCCATTAATTCCATTGAGGCTA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZADH2 PubMed Links

Gene Details

Gene
ZADH2
Gene Name
zinc binding alcohol dehydrogenase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306093.1 7467 UTR 3 NP_001293022.1
NM_175907.5 7467 UTR 3 NP_787103.1

View Full Product Details