Product Details

SNP ID
rs149942920
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:9147914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACTTGGCCTTCAAACAGTTTACC[A/G]TTTTGCTAGAAGAGAAAGGAGATGG
Phenotype
MIM: 610616
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ANKRD12 PubMed Links

Gene Details

Gene
ANKRD12
Gene Name
ankyrin repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083625.2 Intron NP_001077094.1
NM_001204056.1 Intron NP_001190985.1
NM_015208.4 Intron NP_056023.3
XM_005258092.3 Intron XP_005258149.1
XM_005258093.3 Intron XP_005258150.1
XM_005258095.2 Intron XP_005258152.1
XM_005258096.2 Intron XP_005258153.1
XM_011525638.2 Intron XP_011523940.1
XM_011525639.2 Intron XP_011523941.1
XM_017025661.1 Intron XP_016881150.1
XM_017025662.1 Intron XP_016881151.1
XM_017025663.1 Intron XP_016881152.1
XM_017025664.1 Intron XP_016881153.1
XM_017025665.1 Intron XP_016881154.1
XM_017025666.1 Intron XP_016881155.1
XM_017025667.1 Intron XP_016881156.1
XM_017025668.1 Intron XP_016881157.1
XM_017025669.1 Intron XP_016881158.1
XM_017025670.1 Intron XP_016881159.1

View Full Product Details