Product Details
- SNP ID
-
rs150543128
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:46087107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAAGCAGTCAAGGAGCCACCACC[A/G]AAAGCATCGTTCATTTTGGCTGCTC
- Phenotype
-
MIM: 164360
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ATP5A1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8097521] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATP5A1
- Gene Name
- ATP synthase, H+ transporting, mitochondrial F1 complex, alpha subunit 1, cardiac muscle
View Full Product Details