Product Details
- SNP ID
-
rs137866561
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:41837314 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGCTGGTTATGTTCACGAGCACA[C/T]CCCATGAGGAGGAAGGTGGTATTGA
- Phenotype
-
MIM: 614806
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DMRTC2
PubMed Links
Gene Details
- Gene
- DMRTC2
- Gene Name
- DMRT like family C2
There are no transcripts associated with this gene.
- Gene
- LYPD4
- Gene Name
- LY6/PLAUR domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001291419.1 |
814 |
Silent Mutation |
GGA,GGG |
G155G |
NP_001278348.1 |
NM_173506.6 |
814 |
Silent Mutation |
GGA,GGG |
G190G |
NP_775777.3 |
XM_005258543.3 |
814 |
Silent Mutation |
GGA,GGG |
G203G |
XP_005258600.1 |
XM_005278383.4 |
814 |
Silent Mutation |
GGA,GGG |
G203G |
XP_005278440.1 |
XM_005278385.4 |
814 |
Silent Mutation |
GGA,GGG |
G203G |
XP_005278442.1 |
XM_005278387.4 |
814 |
Missense Mutation |
GAT,GGT |
D202G |
XP_005278444.1 |
XM_005278388.3 |
814 |
Silent Mutation |
GGA,GGG |
G141G |
XP_005278445.1 |
XM_006723035.2 |
814 |
Silent Mutation |
GGA,GGG |
G199G |
XP_006723098.1 |
XM_011526483.2 |
814 |
Silent Mutation |
GGA,GGG |
G203G |
XP_011524785.1 |
XM_011526484.2 |
814 |
Silent Mutation |
GGA,GGG |
G203G |
XP_011524786.1 |
View Full Product Details