Product Details

SNP ID
rs137922291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50825886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTGGTGGTGTTGTCACAAGGGAC[A/G]TCACCCCAGGACACAATGCCCTGCA
Phenotype
MIM: 147910 MIM: 610601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK1 PubMed Links

Gene Details

Gene
KLK1
Gene Name
kallikrein 1
There are no transcripts associated with this gene.

Gene
KLK15
Gene Name
kallikrein related peptidase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277081.1 1509 Silent Mutation GAC,GAT D226D NP_001264010.1
NM_001277082.1 1509 UTR 3 NP_001264011.1
NM_017509.3 1509 Silent Mutation GAC,GAT D227D NP_059979.2
XM_006723265.3 1509 Silent Mutation GAC,GAT D227D XP_006723328.1
XM_011527085.2 1509 Silent Mutation GAC,GAT D226D XP_011525387.1
XM_011527087.2 1509 Silent Mutation GAC,GAT D142D XP_011525389.1
XM_011527088.2 1509 UTR 3 XP_011525390.1
XM_011527089.2 1509 UTR 3 XP_011525391.1
XM_017026943.1 1509 Intron XP_016882432.1
Gene
LOC105372441
Gene Name
uncharacterized LOC105372441
There are no transcripts associated with this gene.

Gene
MGC45922
Gene Name
uncharacterized LOC284365
There are no transcripts associated with this gene.

View Full Product Details