Product Details

SNP ID
rs138007892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7639098 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGATATCCTGGCTGAGGGCATCA[C/T]CAGTGAGTGAACGCGTCCCCAGTGA
Phenotype
MIM: 614770 MIM: 601717 MIM: 610850
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCP2 PubMed Links

Gene Details

Gene
PCP2
Gene Name
Purkinje cell protein 2
There are no transcripts associated with this gene.

Gene
PET100
Gene Name
PET100 homolog
There are no transcripts associated with this gene.

Gene
STXBP2
Gene Name
syntaxin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127396.2 216 Missense Mutation ACC,ATC T56I NP_001120868.1
NM_001272034.1 216 Missense Mutation ACC,ATC T56I NP_001258963.1
NM_006949.3 216 Missense Mutation ACC,ATC T56I NP_008880.2
XM_011528210.1 216 Missense Mutation ACC,ATC T56I XP_011526512.1
XM_011528212.2 216 Missense Mutation ACC,ATC T56I XP_011526514.1
Gene
XAB2
Gene Name
XPA binding protein 2
There are no transcripts associated with this gene.

View Full Product Details